COMH4211AAD0 Combi-Blocks, Inc.

3-Bromo-4-(2-methoxy-ethoxy)-benzylamine

  • MDL number MFCD17284188
  • Cas Number : 1342593-68-3
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0987

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0987
Availability details for
Quantity
  • Availability
  • Add to Cart