COMH4211AAD1 Combi-Blocks, Inc.

1-(3-Fluoro-2-isopropoxy-phenyl)-propylamine hydrochloride

  • MDL number MFCD31557791
  • Cas Number : 2206608-22-0
  • Molecular Weight : 247.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0988

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0988
Availability details for
Quantity
  • Availability
  • Add to Cart