COMH4211AAD3 Combi-Blocks, Inc.

5-Bromo-n-cyclopentyl-2-nitroaniline

  • MDL number MFCD21219725
  • Cas Number : 1231930-26-9
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0994

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0994
Availability details for
Quantity
  • Availability
  • Add to Cart