COMH4211AAD4 Combi-Blocks, Inc.

L-Norleucinamide, n-[(phenylmethoxy)carbonyl]-l-alanyl-

  • MDL number MFCD00136681
  • Cas Number : 197237-14-2
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl (1S)-2-{[(1S)-1-(aminocarbonyl)pentyl]amino}-1-methyl-2-oxoethylcarbamate
Inchi Code 1S/C17H25N3O4/c1-3-4-10-14(15(18)21)20-16(22)12(2)19-17(23)24-11-13-8-6-5-7-9-13/h5-9,12,14H,3-4,10-11H2,1-2H3,(H2,18,21)(H,19,23)(H,20,22)/t12-,14-/m0/s1
Inchi Key PTNTUYBFBNVSGD-JSGCOSHPSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0997

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0997
Availability details for
Quantity
  • Availability
  • Add to Cart