COMH4211AAD6 Combi-Blocks, Inc.

Ethyl 3-[(2-propen-1-yloxy)carbonyl]aminobenzoate

  • MDL number MFCD31619026
  • Cas Number : 1160626-00-5
  • Molecular Weight : 249.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1002

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1002
Availability details for
Quantity
  • Availability
  • Add to Cart