COMH4211AAD7 Combi-Blocks, Inc.

1-(3-Isopropoxy-benzyl)-piperazine

  • MDL number MFCD11918591
  • Cas Number : 1181722-00-8
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1003
Availability details for
Quantity
  • Availability
  • Add to Cart