COMH4211AAD8 Combi-Blocks, Inc.

4-Bromo-2-ethynylthiophene

  • MDL number MFCD08703539
  • Cas Number : 690266-87-6
  • Molecular Weight : 187.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1005
Availability details for
Quantity
  • Availability
  • Add to Cart