COMH4211AAD9 Combi-Blocks, Inc.

1-[5-(4-Fluoro-benzyloxy)-2-methoxymethoxy-phenyl]-ethanone

  • MDL number MFCD31618956
  • Cas Number : 500370-53-6
  • Molecular Weight : 304.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1007

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1007
Availability details for
Quantity
  • Availability
  • Add to Cart