COMH4211AADB Combi-Blocks, Inc.

Methyl 5-(3,5-bis(hydroxymethyl)phenoxy)pentanoate

  • MDL number MFCD31614062
  • Cas Number : 1239587-43-9
  • Molecular Weight : 268.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1014
Availability details for
Quantity
  • Availability
  • Add to Cart