COMH4211AADC Combi-Blocks, Inc.

3-Chloro-2-(propan-2-ylsulfanyl)pyridine

  • MDL number MFCD25369036
  • Cas Number : 98626-96-1
  • Molecular Weight : 187.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1015

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1015
Availability details for
Quantity
  • Availability
  • Add to Cart