COMH4211AADE Combi-Blocks, Inc.

tert-Butyl 3,6-dichloropyridine-2-carboxylate

  • MDL number MFCD18897600
  • Cas Number : 211122-64-4
  • Molecular Weight : 248.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1017

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1017
Availability details for
Quantity
  • Availability
  • Add to Cart