COMH4211AAE1 Combi-Blocks, Inc.

4-(2-Hydroxy-1,1-dimethyl-ethylamino)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD29059967
  • Cas Number : 2365419-52-7
  • Molecular Weight : 272.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-((1-hydroxy-2-methylpropan-2-yl)amino)piperidine-1-carboxylate
Inchi Code 1S/C14H28N2O3/c1-13(2,3)19-12(18)16-8-6-11(7-9-16)15-14(4,5)10-17/h11,15,17H,6-10H2,1-5H3
Inchi Key IEEIKKQVECGIOE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1024

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1024
Availability details for
Quantity
  • Availability
  • Add to Cart