COMH4211AAE2 Combi-Blocks, Inc.

4-Ethoxy-2-methoxybenzaldehyde

  • MDL number MFCD24388335
  • Cas Number : 42924-32-3
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1026

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1026
Availability details for
Quantity
  • Availability
  • Add to Cart