COMH4211AAE4 Combi-Blocks, Inc.

1-Bromo-3-(2,2,2-trifluoroethoxy)-5-(trifluoromethyl)benzene

  • MDL number MFCD28400310
  • Cas Number : 1704069-34-0
  • Molecular Weight : 323.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1031
Availability details for
Quantity
  • Availability
  • Add to Cart