COMH4211AAE6 Combi-Blocks, Inc.

1-[3-Bromo-4-(2-methoxy-ethoxy)-phenyl]-ethylamine

  • MDL number MFCD17247748
  • Cas Number : 1339613-48-7
  • Molecular Weight : 274.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1034

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1034
Availability details for
Quantity
  • Availability
  • Add to Cart