COMH4211AAE7 Combi-Blocks, Inc.

5-Amino-4-fluoro-n-isobutyl-2-methyl-benzamide

  • MDL number MFCD31579294
  • Cas Number : 1876008-72-8
  • Molecular Weight : 224.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1038
Availability details for
Quantity
  • Availability
  • Add to Cart