COMH4211AAE9 Combi-Blocks, Inc.

N-(3-Fluorobenzyl)-o-methylhydroxylamine

  • MDL number MFCD18825377
  • Cas Number : 543730-70-7
  • Molecular Weight : 155.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3-fluorobenzyl)-O-methylhydroxylamine
Inchi Code 1S/C8H10FNO/c1-11-10-6-7-3-2-4-8(9)5-7/h2-5,10H,6H2,1H3
Inchi Key UZIVOFAFIGZNQE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1045
Availability details for
Quantity
  • Availability
  • Add to Cart