COMH4211AAEA Combi-Blocks, Inc.

4-(4-Cyclopropylmethoxy-2-fluoro-phenyl)-piperidine hydrochloride

  • MDL number MFCD31583393
  • Cas Number : 2270912-37-1
  • Molecular Weight : 285.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1050

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1050
Availability details for
Quantity
  • Availability
  • Add to Cart