COMH4211AAEB Combi-Blocks, Inc.

Benzeneacetamide, 4-fluoro-n-[4-(5-oxazolyl)phenyl]-

  • MDL number MFCD30486224
  • Cas Number : 1626077-46-0
  • Molecular Weight : 296.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1055
Availability details for
Quantity
  • Availability
  • Add to Cart