COMH4211AAEC Combi-Blocks, Inc.

7-Bromo-5-fluoro-2,3-dihydrobenzo[1,4]dioxine

  • MDL number MFCD30099129
  • Cas Number : 1222556-34-4
  • Molecular Weight : 233.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1059
Availability details for
Quantity
  • Availability
  • Add to Cart