COMH4211AAED Combi-Blocks, Inc.

4-Chloro-3-(2-methoxyethoxy)aniline

  • MDL number MFCD19102114
  • Cas Number : 769961-43-5
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1061
Availability details for
Quantity
  • Availability
  • Add to Cart