COMH4211AAF0 Combi-Blocks, Inc.

N-(Cyclopropylmethyl)-3,5-dimethylaniline

  • MDL number MFCD03211309
  • Cas Number : 356539-49-6
  • Molecular Weight : 175.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1064

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1064
Availability details for
Quantity
  • Availability
  • Add to Cart