COMH4211AAF1 Combi-Blocks, Inc.

1-(4-Ethoxy-2-fluorophenyl)ethanone

  • MDL number MFCD28521106
  • Cas Number : 851667-22-6
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1066
Availability details for
Quantity
  • Availability
  • Add to Cart