COMH4211AAF4 Combi-Blocks, Inc.

1-Ethyl-4-methylpiperidin-4-amine

  • MDL number MFCD12766291
  • Cas Number : 1178743-22-0
  • Molecular Weight : 142.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-ethyl-4-methyl-4-piperidinamine
Inchi Code 1S/C8H18N2/c1-3-10-6-4-8(2,9)5-7-10/h3-7,9H2,1-2H3
Inchi Key ZVRZABLLSMZMOH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1071

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1071
Availability details for
Quantity
  • Availability
  • Add to Cart