COMH4211AAF6 Combi-Blocks, Inc.

(3-Methoxycarbonylmethyl-phenoxy)-acetic acid

  • MDL number MFCD31618878
  • Cas Number : 2270908-04-6
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1076
Availability details for
Quantity
  • Availability
  • Add to Cart