COMH4211AAF7 Combi-Blocks, Inc.

3-(2-Fluoro-5-methylphenoxy)propanoic acid

  • MDL number MFCD20119200
  • Cas Number : 942195-98-4
  • Molecular Weight : 198.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-fluoro-5-methylphenoxy)propanoic acid
Inchi Code 1S/C10H11FO3/c1-7-2-3-8(11)9(6-7)14-5-4-10(12)13/h2-3,6H,4-5H2,1H3,(H,12,13)
Inchi Key YPYVWTSQTHONPY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1080
Availability details for
Quantity
  • Availability
  • Add to Cart