COMH4211AAFA Combi-Blocks, Inc.

Benzyl 9-oxo-3-azaspiro[5.5]undec-7-ene-3-carboxylate

  • MDL number MFCD26743504
  • Cas Number : 189333-18-4
  • Molecular Weight : 299.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1087
Availability details for
Quantity
  • Availability
  • Add to Cart