COMH4211AAFB Combi-Blocks, Inc.

3-(3-Bromo-4-fluoro-phenoxymethyl)-tetrahydro-pyran

  • MDL number MFCD31613872
  • Cas Number : 2270906-06-2
  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1090
Availability details for
Quantity
  • Availability
  • Add to Cart