COMH4211AAFD Combi-Blocks, Inc.

1-(3-Bromophenyl)-3,3-dimethyl-pyrrolidine

  • MDL number MFCD31613892
  • Cas Number : 1334675-31-8
  • Molecular Weight : 254.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1095

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1095
Availability details for
Quantity
  • Availability
  • Add to Cart