COMH4211AB00 Combi-Blocks, Inc.

3-(4,4-Difluoropiperidinyl)propan-1-ol

  • MDL number MFCD30729335
  • Cas Number : 1149751-48-3
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1105
Availability details for
Quantity
  • Availability
  • Add to Cart