COMH4211AB01 Combi-Blocks, Inc.

3-Chloro-4-(4-formylphenoxy)benzonitrile

  • MDL number MFCD31618953
  • Cas Number : 676494-58-9
  • Molecular Weight : 257.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-4-(4-formylphenoxy)benzonitrile
Inchi Code 1S/C14H8ClNO2/c15-13-7-11(8-16)3-6-14(13)18-12-4-1-10(9-17)2-5-12/h1-7,9H
Inchi Key DFOOALCFMCHKGY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1107
Availability details for
Quantity
  • Availability
  • Add to Cart