COMH4211AB02 Combi-Blocks, Inc.

[4-(Cyclobutylmethoxy)phenyl]methanol

  • MDL number MFCD21240222
  • Cas Number : 1224719-91-8
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1108
Availability details for
Quantity
  • Availability
  • Add to Cart