COMH4211AB04 Combi-Blocks, Inc.

Carbamic acid, [1-[[(2,5-dioxo-1-pyrrolidinyl)oxy]carbonyl]propyl]-, 1,1-dimethylethyl ester, (s)-

  • MDL number MFCD26792783
  • Cas Number : 103290-07-9
  • Molecular Weight : 300.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1110

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1110
Availability details for
Quantity
  • Availability
  • Add to Cart