COMH4211AB06 Combi-Blocks, Inc.

N-(4-Bromo-benzyl)-terephthalamic acid

  • MDL number MFCD31614044
  • Cas Number : 1292979-27-1
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1114

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1114
Availability details for
Quantity
  • Availability
  • Add to Cart