COMH4211AB07 Combi-Blocks, Inc.

2-Methyl-4-phenylbutan-1-ol

  • MDL number MFCD20392715
  • Cas Number : 3023-61-8
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1115
Availability details for
Quantity
  • Availability
  • Add to Cart