COMH4211AB08 Combi-Blocks, Inc.

5-Bromo-2-ethoxybenzamide

  • MDL number MFCD07551819
  • Cas Number : 54924-78-6
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1116

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1116
Availability details for
Quantity
  • Availability
  • Add to Cart