COMH4211AB09 Combi-Blocks, Inc.

5-Bromo-3,4-dichloro-n,n-bis(4-methoxybenzyl)pyridin-2-amine

  • MDL number MFCD30608835
  • Cas Number : 1607838-03-8
  • Molecular Weight : 482.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1118

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1118
Availability details for
Quantity
  • Availability
  • Add to Cart