COMH4211AB0A Combi-Blocks, Inc.

2-Amino-6-chloro-4-[(3-hydroxypropyl)amino]pyrimidine hydrochloride

  • MDL number MFCD31618983
  • Cas Number : 199453-07-1
  • Molecular Weight : 239.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1122

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1122
Availability details for
Quantity
  • Availability
  • Add to Cart