COMH4211AB0B Combi-Blocks, Inc.

Thiomorpholine, 4-[(4-bromo-2-fluorophenyl)methyl]-, 1,1-dioxide

  • MDL number MFCD16197188
  • Cas Number : 1092563-28-4
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1124

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1124
Availability details for
Quantity
  • Availability
  • Add to Cart