COMH4211AB0D Combi-Blocks, Inc.

2,5-Dichloro-n-methylnicotinamide

  • MDL number MFCD18642436
  • Cas Number : 931105-53-2
  • Molecular Weight : 205.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-dichloro-N-methylnicotinamide
Inchi Code 1S/C7H6Cl2N2O/c1-10-7(12)5-2-4(8)3-11-6(5)9/h2-3H,1H3,(H,10,12)
Inchi Key VPMWKZHRYGXBCK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1128

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1128
Availability details for
Quantity
  • Availability
  • Add to Cart