COMH4211AB0E Combi-Blocks, Inc.

6-Chloro-n-(cyclopropylmethyl)pyridine-3-carboxamide

  • MDL number MFCD09805007
  • Cas Number : 585544-03-2
  • Molecular Weight : 210.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1129

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1129
Availability details for
Quantity
  • Availability
  • Add to Cart