COMH4211AB10 Combi-Blocks, Inc.

2-Bromo-5-ethoxy-4-methyl-benzylamine hydrochloride

  • MDL number MFCD31557811
  • Cas Number : 2203940-76-3
  • Molecular Weight : 280.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1132
Availability details for
Quantity
  • Availability
  • Add to Cart