COMH4211AB11 Combi-Blocks, Inc.

1-(4-Bromobutoxy)-2-chlorobenzene

  • MDL number MFCD02030538
  • Cas Number : 23468-00-0
  • Molecular Weight : 263.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1133

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1133
Availability details for
Quantity
  • Availability
  • Add to Cart