COMH4211AB12 Combi-Blocks, Inc.

Trans-4-[n-methyl-n-(tert-butoxycarbonyl)-amino]cyclohexanecarboxylic acid

  • MDL number MFCD28383642
  • Cas Number : 609805-41-6
  • Molecular Weight : 257.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1140

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1140
Availability details for
Quantity
  • Availability
  • Add to Cart