COMH4211AB14 Combi-Blocks, Inc.

1-[3-(2-Methoxy-ethoxy)-phenyl]-ethylamine hydrochloride

  • MDL number MFCD31381724
  • Cas Number : 2206606-55-3
  • Molecular Weight : 231.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1143

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1143
Availability details for
Quantity
  • Availability
  • Add to Cart