COMH4211AB16 Combi-Blocks, Inc.

Toluene-4-sulfonic acid 2-(3-trityloxy-propoxy)-ethyl ester

  • MDL number MFCD31618938
  • Cas Number : 1020728-53-3
  • Molecular Weight : 516.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1150
Availability details for
Quantity
  • Availability
  • Add to Cart