COMH4211AB18 Combi-Blocks, Inc.

Dimethyl (6-chloropyrimidin-4-yl)carbonimidodithioate

  • MDL number MFCD30530612
  • Cas Number : 1192814-58-6
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1153
Availability details for
Quantity
  • Availability
  • Add to Cart