COMH4211AB1A Combi-Blocks, Inc.

4-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenylmethanesulfonyl]-morpholine

  • MDL number MFCD31579501
  • Cas Number : 1620955-85-2
  • Molecular Weight : 367.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1159
Availability details for
Quantity
  • Availability
  • Add to Cart