COMH4211AB1B Combi-Blocks, Inc.

1-(4-Bromo-2-cyclopropylmethoxy-phenyl)-ethanone

  • MDL number MFCD24110650
  • Cas Number : 1548886-22-1
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1160

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1160
Availability details for
Quantity
  • Availability
  • Add to Cart