COMH4211AB1C Combi-Blocks, Inc.

2-[3-Chloro-4-(cyclopropylmethoxy)phenyl]acetic acid

  • MDL number MFCD30580986
  • Cas Number : 57017-96-6
  • Molecular Weight : 240.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1161

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1161
Availability details for
Quantity
  • Availability
  • Add to Cart